2-Amino-N-cyclopropylbenzamide

Supplier: Matrix Scientific

061154-500MG
101921-408EA 272.55 USD
101921-408
2-Amino-N-cyclopropylbenzamide
2-Amino-N-cyclopropylbenzamide

Hazard: Irritant

Formula: C₁₀H₁₂N₂O
MDL Number: MFCD03015446
CAS Number: 30510-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR