3-[4-(Pyrrolidin-1-ylsulfonyl)phenyl]propanoic acid

Supplier: Matrix Scientific

061152-500MG
101921-404EA 364.62 USD
101921-404
3-[4-(Pyrrolidin-1-ylsulfonyl)phenyl]propanoic acid
3-[4-(Pyrrolidin-1-ylsulfonyl)phenyl]propanoic acid

Hazard: Irritant

Formula: C₁₃H₁₇NO₄S
MDL Number: MFCD07352319
CAS Number: 871544-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR