(1-Adamantylthio)acetic acid

Supplier: Matrix Scientific

061149-500MG
101921-398EA 183.08 USD
101921-398
(1-Adamantylthio)acetic acid
(1-Adamantylthio)acetic acid

Hazard: Irritant

Formula: C₁₂H₁₈O₂S
MDL Number: MFCD01821191
CAS Number: 95769-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR