6-Ethoxy-2,2,4-trimethyl-1,2,3,4-tetrahydroquinoline

Supplier: Matrix Scientific

061148-500MG
101921-396EA 484.62 USD
101921-396
6-Ethoxy-2,2,4-trimethyl-1,2,3,4-tetrahydroquinoline
6-Ethoxy-2,2,4-trimethyl-1,2,3,4-tetrahydroquinoline

Hazard: Irritant

Formula: C₁₄H₂₁NO
MDL Number: MFCD00006694
CAS Number: 16489-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR