[3,5-Dimethyl-4-(2-oxo-2-pyrrolidin-1-ylethoxy)phenyl]amine

Supplier: Matrix Scientific

061144-500MG
101921-388EA 364.62 USD
101921-388
[3,5-Dimethyl-4-(2-oxo-2-pyrrolidin-1-ylethoxy)phenyl]amine
[3,5-Dimethyl-4-(2-oxo-2-pyrrolidin-1-ylethoxy)phenyl]amine

Hazard: Irritant

Formula: C₁₄H₂₀N₂O₂
MDL Number: MFCD07658186
CAS Number: 893766-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR