3-Amino-N-cyclopropylbenzenesulfonamide

Supplier: Matrix Scientific

061141-500MG
101921-382EA 50.77 USD
101921-382
3-Amino-N-cyclopropylbenzenesulfonamide
3-Amino-N-cyclopropylbenzenesulphonamide

Hazard: Irritant/Refrigerate

Formula: C₉H₁₂N₂O₂S
MDL Number: MFCD07022080
CAS Number: 459434-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR