2-Chloro-N-[(3-oxo-3,4-dihydro-2H-1,4-benzoxazin-6-yl)methyl]acetamide
Supplier: Matrix Scientific
061140-500MG
101921-380EA
364.62
USD
101921-380
2-Chloro-N-[(3-oxo-3,4-dihydro-2H-1,4-benzoxazin-6-yl)methyl]acetamide
2-Chloro-N-[(3-oxo-3,4-dihydro-2H-1,4-benzoxazin-6-yl)methyl]acetamide
Hazard: Irritant
Formula:
C₁₁H₁₁ClN₂O₃ |
MDL Number:
MFCD03196010 CAS Number: 540512-38-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...