4-Amino-3,5-diethyl-1,2,4-triazole

Supplier: Matrix Scientific

061132-500MG
101921-364EA 484.62 USD
101921-364
4-Amino-3,5-diethyl-1,2,4-triazole
4-Amino-3,5-diethyl-1,2,4-triazole

Hazard: Irritant

Formula: C₆H₁₂N₄
CAS Number: 6285-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR