1-(2-Fluorobenzyl)-1H-indole-2,3-dione
Supplier: Matrix Scientific
061128-500MG
101921-356EA
243.08
USD
101921-356
1-(2-Fluorobenzyl)-1H-indole-2,3-dione
1-(2-Fluorobenzyl)-1H-indole-2,3-dione
Hazard: Irritant
Formula:
C₁₅H₁₀FNO₂ |
MDL Number:
MFCD01907030 CAS Number: 346640-52-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...