[4-(2-Morpholin-4-yl-2-oxoethoxy)phenyl]amine

Supplier: Matrix Scientific

061126-500MG
101921-352EA 606.15 USD
101921-352
[4-(2-Morpholin-4-yl-2-oxoethoxy)phenyl]amine
[4-(2-Morpholin-4-yl-2-oxoethoxy)phenyl]amine

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₃
MDL Number: MFCD01209797
CAS Number: 76870-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR