5-Chloro-1-(3-chlorophenyl)-3-methyl-1H-pyrazole-4-carbaldehyde

Supplier: Matrix Scientific

061124-500MG
101921-348EA 484.62 USD
101921-348
5-Chloro-1-(3-chlorophenyl)-3-methyl-1H-pyrazole-4-carbaldehyde
5-Chloro-1-(3-chlorophenyl)-3-methyl-1H-pyrazole-4-carbaldehyde

Hazard: Irritant

Formula: C₁₁H₈Cl₂N₂O
MDL Number: MFCD03980997
CAS Number: 77509-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR