[4-(3-Methyl-1H-pyrazol-1-yl)phenyl]amine

Supplier: Matrix Scientific

061122-500MG
101921-344EA 606.15 USD
101921-344
[4-(3-Methyl-1H-pyrazol-1-yl)phenyl]amine
[4-(3-Methyl-1H-pyrazol-1-yl)phenyl]amine

Hazard: Irritant

Formula: C₁₀H₁₁N₃
MDL Number: MFCD03768864
CAS Number: 53006-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR