Methyl-3-amino-4-piperidin-1-ylbenzoate

Supplier: Matrix Scientific

061121-500MG
101921-342EA 183.08 USD
101921-342
Methyl-3-amino-4-piperidin-1-ylbenzoate
Methyl-3-amino-4-piperidin-1-ylbenzoate

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₂
MDL Number: MFCD05257068
CAS Number: 696616-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR