(4-Aminophenyl)(2-thienyl)methanone

Supplier: Matrix Scientific

061117-500MG
101921-334EA 364.62 USD
101921-334
(4-Aminophenyl)(2-thienyl)methanone
(4-Aminophenyl)(2-thienyl)methanone

Hazard: Irritant

Formula: C₁₁H₉NOS
MDL Number: MFCD03368394
CAS Number: 25706-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR