5-Oxo-2,3-dihydro-1H,5H-pyrido[3,2,1-ij]quinoline-7-carbaldehyde

Supplier: Matrix Scientific

061116-500MG
101921-332EA 243.08 USD
101921-332
5-Oxo-2,3-dihydro-1H,5H-pyrido[3,2,1-ij]quinoline-7-carbaldehyde
5-Oxo-2,3-dihydro-1H,5H-pyrido[3,2,1-ij]quinoline-7-carbaldehyde

Hazard: Irritant

Formula: C₁₃H₁₁NO₂
MDL Number: MFCD03198261
CAS Number: 111724-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR