Dimethyl-5-(5-formyl-2-furyl)isophthalate

Supplier: Matrix Scientific

061105-500MG
101921-310EA 243.08 USD
101921-310
Dimethyl-5-(5-formyl-2-furyl)isophthalate
Dimethyl-5-(5-formyl-2-furyl)isophthalate

Hazard: Irritant

Formula: C₁₅H₁₂O₆
MDL Number: MFCD03102982
CAS Number: 591226-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR