5-Hydroxy-3-methyl-1-(4-methylphenyl)-1H-pyrazole-4-carbaldehyde

Supplier: Matrix Scientific

061104-500MG
101921-308EA 484.62 USD
101921-308
5-Hydroxy-3-methyl-1-(4-methylphenyl)-1H-pyrazole-4-carbaldehyde
5-Hydroxy-3-methyl-1-(4-methylphenyl)-1H-pyrazole-4-carbaldehyde

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₂
MDL Number: MFCD12197689
CAS Number: 60484-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR