2-Aminoquinoline-3-carbonitrile

Supplier: Matrix Scientific

061089-500MG
101921-278EA 484.62 USD
101921-278
2-Aminoquinoline-3-carbonitrile
2-Aminoquinoline-3-carbonitrile

Hazard: Irritant

Formula: C₁₀H₇N₃
MDL Number: MFCD00662531
CAS Number: 31407-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR