5-(1-Adamantyl)-2-furohydrazide

Supplier: Matrix Scientific

061087-500MG
101921-274EA 397.9 USD
101921-274
5-(1-Adamantyl)-2-furohydrazide
5-(1-Adamantyl)-2-furohydrazide

Hazard: Irritant

Formula: C₁₅H₂₀N₂O₂
MDL Number: MFCD07626327
CAS Number: 897545-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR