Adamantane-1-carbohydrazide

Supplier: Matrix Scientific

061084-500MG
101921-268EA 273.85 USD
101921-268
Adamantane-1-carbohydrazide
Adamantane-1-carbohydrazide

Hazard: Irritant

Formula: C₁₁H₁₈N₂O
MDL Number: MFCD00154679
CAS Number: 17846-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR