1-Butyl-1H-indole-2,3-dione
Supplier: Matrix Scientific
061083-500MG
101921-266EA
243.08
USD
101921-266
1-Butyl-1H-indole-2,3-dione
1-Butyl-1H-indole-2,3-dione
Hazard: Irritant
Formula:
C₁₂H₁₃NO₂ |
MDL Number:
MFCD00224221 CAS Number: 4290-91-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...