2-Amino-N-(3-chlorophenyl)quinoline-3-carboxamide

Supplier: Matrix Scientific

061079-500MG
101921-258EA 290.77 USD
101921-258
2-Amino-N-(3-chlorophenyl)quinoline-3-carboxamide
2-Amino-N-(3-chlorophenyl)quinoline-3-carboxamide

Hazard: Irritant

Formula: C₁₆H₁₂ClN₃O
MDL Number: MFCD03134092
CAS Number: 446052-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR