5-Oxo-1-(2-phenoxyethyl)pyrrolidine-3-carboxylic acid

Supplier: Matrix Scientific

061077-500MG
101921-254EA 364.62 USD
101921-254
5-Oxo-1-(2-phenoxyethyl)pyrrolidine-3-carboxylic acid
5-Oxo-1-(2-phenoxyethyl)pyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₅NO₄
MDL Number: MFCD02625749
CAS Number: 361985-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR