Ethyl-5-(4-methoxyphenyl)-7-(trifluoromethyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine-3-carboxylate

Supplier: Matrix Scientific

061075-500MG
101921-250EA 364.62 USD
101921-250
Ethyl-5-(4-methoxyphenyl)-7-(trifluoromethyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine-3-carboxylate
Ethyl-5-(4-methoxyphenyl)-7-(trifluoromethyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine-3-carboxylate

Hazard: Irritant

Formula: C₁₇H₁₈F₃N₃O₃
MDL Number: MFCD04049064
CAS Number: 667920-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR