Ethyl-5-(4-chlorophenyl)-7-(trifluoromethyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine-3-carboxylate

Supplier: Matrix Scientific

061074-500MG
101921-248EA 364.62 USD
101921-248
Ethyl-5-(4-chlorophenyl)-7-(trifluoromethyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine-3-carboxylate
Ethyl-5-(4-chlorophenyl)-7-(trifluoromethyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine-3-carboxylate

Hazard: Irritant

Formula: C₁₆H₁₅ClF₃N₃O₂
MDL Number: MFCD04048251
CAS Number: 667920-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR