3-Methoxy-5-(1H-tetrazol-1-yl)aniline

Supplier: Matrix Scientific

061057-500MG
101921-216EA 364.62 USD
101921-216
3-Methoxy-5-(1H-tetrazol-1-yl)aniline
3-Methoxy-5-(1H-tetrazol-1-yl)aniline

Hazard: Irritant

Formula: C₈H₉N₅O
MDL Number: MFCD03419552
CAS Number: 883291-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR