1-Ethyl-2-pyrrolidin-2-yl-1H-benzimidazole dihydrochloride

Supplier: Matrix Scientific

061051-500MG
101921-204EA 484.62 USD
101921-204
1-Ethyl-2-pyrrolidin-2-yl-1H-benzimidazole dihydrochloride
1-Ethyl-2-pyrrolidin-2-yl-1H-benzimidazole dihydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉Cl₂N₃
MDL Number: MFCD19103653
CAS Number: 1332530-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR