N-(2-Chloroethyl)-1H-benzimidazol-2-amine hydrochloride

Supplier: Matrix Scientific

061047-500MG
101921-196EA 364.62 USD
101921-196
N-(2-Chloroethyl)-1H-benzimidazol-2-amine hydrochloride
N-(2-Chloroethyl)-1H-benzimidazol-2-amine hydrochloride

Hazard: Irritant

Formula: C₉H₁₁Cl₂N₃
MDL Number: MFCD16606014
CAS Number: 84797-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR