1-Oxa-9-azaspiro[5.5]undecan-4-ol hydrochloride

Supplier: Matrix Scientific

061044-500MG
101921-190EA 727.69 USD
101921-190
1-Oxa-9-azaspiro[5.5]undecan-4-ol hydrochloride
1-Oxa-9-azaspiro[5.5]undecan-4-ol hydrochloride

Hazard: Irritant

Formula: C₉H₁₈ClNO₂
MDL Number: MFCD19103508
CAS Number: 1332529-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR