[2-(5-Cyclopentyl-1,2,4-oxadiazol-3-yl)ethyl]amine hydrochloride

Supplier: Matrix Scientific

061030-500MG
101921-162EA 548.55 USD
101921-162
[2-(5-Cyclopentyl-1,2,4-oxadiazol-3-yl)ethyl]amine hydrochloride
[2-(5-Cyclopentyl-1,2,4-oxadiazol-3-yl)ethyl]amine hydrochloride

Hazard: Irritant

Formula: C₉H₁₆ClN₃O
MDL Number: MFCD19103490
CAS Number: 1244803-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR