2-(Piperidin-4-yloxy)benzoic acid hydrochloride

Supplier: Matrix Scientific

061025-500MG
101921-152EA 223.08 USD
101921-152
2-(Piperidin-4-yloxy)benzoic acid hydrochloride
2-(Piperidin-4-yloxy)benzoic acid hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₆ClNO₃
MDL Number: MFCD19103486
CAS Number: 1243250-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR