3-(4-Benzyl-4H-1,2,4-triazol-3-yl)piperidine dihydrochloride
Supplier: Matrix Scientific
061018-500MG
101921-138EA
364.62
USD
101921-138
3-(4-Benzyl-4H-1,2,4-triazol-3-yl)piperidine dihydrochloride
3-(4-Benzyl-4H-1,2,4-triazol-3-yl)piperidine dihydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₀Cl₂N₄ |
MDL Number:
MFCD19103478 CAS Number: 1332528-92-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...