3-Propylpiperidine-3-carboxylic acid hydrochloride

Supplier: Matrix Scientific

061017-500MG
101921-136EA 364.62 USD
101921-136
3-Propylpiperidine-3-carboxylic acid hydrochloride
3-Propylpiperidine-3-carboxylic acid hydrochloride

Hazard: Irritant

Formula: C₉H₁₈ClNO₂
MDL Number: MFCD19103477
CAS Number: 1332529-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR