4-(4-Isopropyl-4H-1,2,4-triazol-3-yl)piperidine dihydrochloride

Supplier: Matrix Scientific

061016-500MG
101921-134EA 602.6 USD
101921-134
4-(4-Isopropyl-4H-1,2,4-triazol-3-yl)piperidine dihydrochloride
4-(4-Isopropyl-4H-1,2,4-triazol-3-yl)piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₀Cl₂N₄
MDL Number: MFCD18089418
CAS Number: 1305712-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR