3-(5-Cyclopropyl-4H-1,2,4-triazol-3-yl)piperidine dihydrochloride

Supplier: Matrix Scientific

061012-500MG
101921-126EA 338.46 USD
101921-126
3-(5-Cyclopropyl-4H-1,2,4-triazol-3-yl)piperidine dihydrochloride
3-(5-Cyclopropyl-4H-1,2,4-triazol-3-yl)piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₀H₁₈Cl₂N₄
MDL Number: MFCD19103472
CAS Number: 1332530-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR