Methyl-2-propylpiperidine-2-carboxylate hydrochloride

Supplier: Matrix Scientific

061009-500MG
101921-120EA 364.62 USD
101921-120
Methyl-2-propylpiperidine-2-carboxylate hydrochloride
Methyl-2-propylpiperidine-2-carboxylate hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₀ClNO₂
MDL Number: MFCD19103471
CAS Number: 1332529-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR