2-Methylpiperidine-2-carboxylic acid hydrochloride

Supplier: Matrix Scientific

061007-500MG
101921-116EA 606.15 USD
101921-116
2-Methylpiperidine-2-carboxylic acid hydrochloride
2-Methylpiperidine-2-carboxylic acid hydrochloride

Hazard: Irritant

Formula: C₇H₁₃NO₂·ClH
MW: 179.645 g/mol
MDL Number: MFCD19103470
CAS Number: 72518-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR