Methyl-4-ethylpiperidine-4-carboxylate hydrochloride

Supplier: Matrix Scientific

061006-500MG
101921-114EA 172.31 USD
101921-114
Methyl-4-ethylpiperidine-4-carboxylate hydrochloride
Methyl-4-ethylpiperidine-4-carboxylate hydrochloride

Hazard: Irritant

Formula: C₉H₁₈ClNO₂
MDL Number: MFCD12408565
CAS Number: 1206228-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR