3-Ethylpiperidine-3-carboxylic acid hydrochloride

Supplier: Matrix Scientific

061002-500MG
101921-106EA 522.1 USD
101921-106
3-Ethylpiperidine-3-carboxylic acid hydrochloride
3-Ethylpiperidine-3-carboxylic acid hydrochloride

Hazard: Irritant

Formula: C₈H₁₆ClNO₂
MDL Number: MFCD19103467
CAS Number: 116140-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR