4-[4-(Piperidin-3-ylmethoxy)benzoyl]morpholine hydrochloride

Supplier: Matrix Scientific

060989-500MG
101921-080EA 243.08 USD
101921-080
4-[4-(Piperidin-3-ylmethoxy)benzoyl]morpholine hydrochloride
4-[4-(Piperidin-3-ylmethoxy)benzoyl]morpholine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₅ClN₂O₃
MDL Number: MFCD19103455
CAS Number: 1332529-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR