3-(3-Benzyl-1,2,4-oxadiazol-5-yl)-1-methyl-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine HCl

Supplier: Matrix Scientific

060988-500MG
101921-078EA 290.77 USD
101921-078
3-(3-Benzyl-1,2,4-oxadiazol-5-yl)-1-methyl-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine HCl
3-(3-Benzyl-1,2,4-oxadiazol-5-yl)-1-methyl-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine HCl

Hazard: Irritant

Formula: C₁₆H₁₈ClN₅O
MDL Number: MFCD19103454
CAS Number: 1332530-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR