1-Methyl-3-(3-phenyl-1,2,4-oxadiazol-5-yl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine HCl

Supplier: Matrix Scientific

060983-500MG
101921-068EA 364.62 USD
101921-068
1-Methyl-3-(3-phenyl-1,2,4-oxadiazol-5-yl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine HCl
1-Methyl-3-(3-phenyl-1,2,4-oxadiazol-5-yl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine HCl

Hazard: Irritant

Formula: C₁₅H₁₆ClN₅O
MDL Number: MFCD19103449
CAS Number: 1332528-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR