N-Cyclopropyl-4-(piperidin-3-ylmethoxy)benzamide hydrochloride

Supplier: Matrix Scientific

060974-500MG
101921-050EA 196.92 USD
101921-050
N-Cyclopropyl-4-(piperidin-3-ylmethoxy)benzamide hydrochloride
N-Cyclopropyl-4-(piperidin-3-ylmethoxy)benzamide hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₃ClN₂O₂
MDL Number: MFCD19103439
CAS Number: 1332530-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR