4-Methyl-1-[4-(piperidin-4-ylmethoxy)benzoyl]piperidine hydrochloride

Supplier: Matrix Scientific

060973-500MG
101921-048EA 243.08 USD
101921-048
4-Methyl-1-[4-(piperidin-4-ylmethoxy)benzoyl]piperidine hydrochloride
4-Methyl-1-[4-(piperidin-4-ylmethoxy)benzoyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₉H₂₉ClN₂O₂
MW: 352.91 g/mol
MDL Number: MFCD19103438
CAS Number: 1332531-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR