Ethyl-5-[(cyclohexylamino)methyl]-2,4-dimethyl-1H-pyrrole-3-carboxylate hydrochloride
Supplier: Matrix Scientific
060960-500MG
101921-022EA
193.85
USD
101921-022
Ethyl-5-[(cyclohexylamino)methyl]-2,4-dimethyl-1H-pyrrole-3-carboxylate hydrochloride
Ethyl-5-[(cyclohexylamino)methyl]-2,4-dimethyl-1H-pyrrole-3-carboxylate hydrochloride
Hazard: Irritant
Formula:
C₁₆H₂₇ClN₂O₂ |
MDL Number:
MFCD19103425 CAS Number: 859067-14-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...