5-Fluoro-2-piperidin-2-yl-1H-benzimidazole dihydrochloride

Supplier: Matrix Scientific

060951-500MG
101921-004EA 606.15 USD
101921-004
5-Fluoro-2-piperidin-2-yl-1H-benzimidazole dihydrochloride
5-Fluoro-2-piperidin-2-yl-1H-benzimidazole dihydrochloride

Hazard: Irritant

Formula: C₁₂H₁₆Cl₂FN₃
MDL Number: MFCD11841200
CAS Number: 885275-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR