3-Amino-1-methyl-7-(trifluoromethyl)-1,3-dihydro-2H-indol-2-one hydrochloride

Supplier: Matrix Scientific

060947-500MG
101920-996EA 364.62 USD
101920-996
3-Amino-1-methyl-7-(trifluoromethyl)-1,3-dihydro-2H-indol-2-one hydrochloride
3-Amino-1-methyl-7-(trifluoromethyl)-1,3-dihydro-2H-indol-2-one hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₀ClF₃N₂O
MDL Number: MFCD19103380
CAS Number: 1332528-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR