3-(Methylthio)propanimidamide hydrochloride

Supplier: Matrix Scientific

060944-500MG
101920-992EA 364.62 USD
101920-992
3-(Methylthio)propanimidamide hydrochloride
3-(Methylthio)propanimidamide hydrochloride

Hazard: Irritant

Formula: C₄H₁₁ClN₂S
MDL Number: MFCD18380688
CAS Number: 88570-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR