2-(2-Thienyl)ethanimidamide hydrochloride

Supplier: Matrix Scientific

060943-500MG
101920-990EA 693.45 USD
101920-990
2-(2-Thienyl)ethanimidamide hydrochloride
2-(2-Thienyl)ethanimidamide hydrochloride

Hazard: Irritant

Formula: C₆H₉ClN₂S
MDL Number: MFCD08752299
CAS Number: 6449-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR