4-(Chloromethyl)-1-ethyl-1H-pyrazole hydrochloride

Supplier: Matrix Scientific

060929-500MG
101920-962EA 469.23 USD
101920-962
4-(Chloromethyl)-1-ethyl-1H-pyrazole hydrochloride
4-(Chloromethyl)-1-ethyl-1H-pyrazole hydrochloride

Hazard: Irritant

Formula: C₆H₁₀Cl₂N₂
MDL Number: MFCD19104828
CAS Number: 1332529-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR